siRNA Details
| Gene Symbol: Sp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013672.2 | NP_038700.2 | 3 | 385 | ||
| GenBank mRNA | AF022363.1 | - | 3 | 373 | ||
| AF062566.1 | - | 3 | 321 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sp1| Gene Symbol: Sp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013672.2 | NP_038700.2 | 3 | 385 | ||
| GenBank mRNA | AF022363.1 | - | 3 | 373 | ||
| AF062566.1 | - | 3 | 321 | |||