siRNA Details
| Gene Symbol: Thbs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013691.2 | NP_038719.2 | 1, 2 | 136 | ||
| GenBank mRNA | BC053023.1 | - | 1, 2 | 90 | 6401682 | |
| BC060104.1 | - | 1, 2 | 96 | |||
| CJ064850.1 | - | 1, 2 | 137 | |||
| L04302.1 | - | 1, 2 | 116 | |||
| L24434.1 | - | 1, 2 | 113 | |||

Thbs3
Genomic Map