siRNA Details
| Gene Symbol: Zfp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013705.1 | NP_038733.1 | 3, 4 | 335 | ||
| GenBank mRNA | AK014064.1 | - | 2, 3 | 177 | ||
| AY605122.1 | - | 3, 4 | 335 | |||
| BC049120.1 | - | 3, 4 | 336 | 6813126 | ||
| BC051972.1 | - | 3, 4 | 535 | 5707652 | ||
| Z30174.1 | - | 3, 4 | 335 | |||

Zfp30
Genomic Map