siRNA Details
| Gene Symbol: CHRNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000748.2 | NP_000739.1 | 6 | 2043 | ||
| XM_017000180.1 | XP_016855669.1 | 6 | 1889 | |||
| GenBank mRNA | U62437.1 | - | 6 | 2043 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHRNB2| Gene Symbol: CHRNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000748.2 | NP_000739.1 | 6 | 2043 | ||
| XM_017000180.1 | XP_016855669.1 | 6 | 1889 | |||
| GenBank mRNA | U62437.1 | - | 6 | 2043 | ||