siRNA Details
| Gene Symbol: Usp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013700.1 | NP_038728.1 | 4 | 414 | ||
| GenBank mRNA | AK154008.1 | - | 4 | 403 | ||
| AK184039.1 | - | 3 | 221 | |||
| AK213919.1 | - | 1 | 71 | |||
| AK219195.1 | - | 1 | 71 | |||
| BC066993.1 | - | 4 | 392 | 5689229 | ||

Usp5
Genomic Map