siRNA Details
| Gene Symbol: Zfp30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013705.1 | NP_038733.1 | 4 | 385 | ||
| GenBank mRNA | AK014064.1 | - | 3 | 227 | ||
| AK030562.1 | - | 1193 | ||||
| AK032058.1 | - | 963 | ||||
| AK173073.1 | - | 1001 | ||||
| AY605122.1 | - | 4 | 385 | |||
| BC049120.1 | - | 4 | 386 | 6813126 | ||
| BC051972.1 | - | 4 | 585 | 5707652 | ||
| Z30174.1 | - | 4 | 385 | |||

Zfp30
Genomic Map