siRNA Details
| Gene Symbol: Mga | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164274.1 | NP_001157746.1 | 2 | 494 | ||
| NM_013720.2 | NP_038748.2 | 2 | 494 | |||
| GenBank mRNA | AF205935.1 | - | 1 | 308 | ||
| AK012050.1 | - | 2 | 457 | |||
| AK032570.1 | - | 2 | 475 | |||
| AK046252.1 | - | 2 | 495 | |||
| AK133775.1 | - | 2 | 711 | |||
| AK139919.1 | - | 2 | 446 | |||
| AK145491.1 | - | 2 | 443 | |||
| AK162302.1 | - | 2 | 491 | |||
| AK172962.1 | - | 2 | 417 | |||

Mga
Genomic Map