siRNA Details
| Gene Symbol: Psmb10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013640.3 | NP_038668.2 | 8 | 1223 | ||
| GenBank mRNA | AK144096.1 | - | 8 | 946 | ||
| BC004730.1 | - | 8 | 949 | 3583052 | ||
| D85561.1 | - | 8 | 918 | |||
| U77784.1 | - | 8 | 910 | |||
| Y10875.1 | - | 8 | 941 | |||

Psmb10
Genomic Map