siRNA Details
| Gene Symbol: Surf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013678.2 | NP_038706.1 | 4 | 380 | ||
| GenBank mRNA | AK004931.1 | - | 1 | 2193 | ||
| AK157125.1 | - | 4 | 394 | |||
| AK165559.1 | - | 4 | 387 | |||
| AK166181.1 | - | 4 | 409 | |||
| AK172018.1 | - | 4 | 383 | |||
| AK177886.1 | - | 57 | ||||
| AK188020.1 | - | 2 | 57 | |||
| BC049779.1 | - | 3 | 1711 | |||
| BC096401.1 | - | 4 | 380 | 1365532 | ||

Surf2
Genomic Map