siRNA Details
| Gene Symbol: GRIN2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000836.2 | NP_000827.2 | 6 | 1551 | ||
| XM_011526872.1 | XP_011525174.1 | 5 | 1719 | |||
| GenBank mRNA | U77783.1 | - | 6 | 1551 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRIN2D| Gene Symbol: GRIN2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000836.2 | NP_000827.2 | 6 | 1551 | ||
| XM_011526872.1 | XP_011525174.1 | 5 | 1719 | |||
| GenBank mRNA | U77783.1 | - | 6 | 1551 | ||