siRNA Details
| Gene Symbol: Krtap15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013713.1 | NP_038741.1 | 1 | 794 | ||
| GenBank mRNA | AK132534.1 | - | 1 | 810 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Krtap15| Gene Symbol: Krtap15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013713.1 | NP_038741.1 | 1 | 794 | ||
| GenBank mRNA | AK132534.1 | - | 1 | 810 | ||