siRNA Details
| Gene Symbol: Eif2ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177806.1 | NP_001171277.1 | 38 | 4780 | ||
| NM_013719.3 | NP_038747.2 | 39 | 5084 | |||
| GenBank mRNA | AJ243533.1 | - | 39 | 5084 | ||
| AK011380.1 | - | 12 | 1170 | |||
| AK129334.1 | - | 34 | 4434 | |||
| BC072637.1 | - | 38 | 4780 | 30354433 | ||

Eif2ak4
Genomic Map