siRNA Details
| Gene Symbol: Olfr154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013728.2 | NP_038756.2 | 1 | 331 | ||
| GenBank mRNA | BC120642.1 | - | 1 | 331 | 40129565 | |
| BC120644.1 | - | 1 | 331 | 40129567 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr154| Gene Symbol: Olfr154 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013728.2 | NP_038756.2 | 1 | 331 | ||
| GenBank mRNA | BC120642.1 | - | 1 | 331 | 40129565 | |
| BC120644.1 | - | 1 | 331 | 40129567 | ||