siRNA Details
| Gene Symbol: Csnk1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289898.1 | NP_001276827.1 | 3 | 245 | ||
| NM_001289899.1 | NP_001276828.1 | 3 | 717 | |||
| NM_013767.6 | NP_038795.3 | 3 | 249 | |||
| GenBank mRNA | AB028241.1 | - | 3 | 195 | ||
| AB028736.2 | - | 3 | 195 | |||
| AK158709.1 | - | 3 | 718 | |||
| BC026127.1 | - | 3 | 198 | 4010696 | ||

Csnk1e
Genomic Map