siRNA Details
| Gene Symbol: Tcl1b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013776.1 | NP_038804.1 | 3 | 422 | ||
| GenBank mRNA | AF195493.1 | - | 3 | 422 | ||
| AK139627.1 | - | 3 | 437 | |||
| BC125294.1 | - | 3 | 419 | 40129809 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tcl1b5| Gene Symbol: Tcl1b5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013776.1 | NP_038804.1 | 3 | 422 | ||
| GenBank mRNA | AF195493.1 | - | 3 | 422 | ||
| AK139627.1 | - | 3 | 437 | |||
| BC125294.1 | - | 3 | 419 | 40129809 | ||