siRNA Details
| Gene Symbol: Ip6k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013785.2 | NP_038813.2 | 2, 3 | 531 | ||
| GenBank mRNA | AF177144.1 | - | 1, 2 | 231 | ||
| AK088872.1 | - | 2, 3 | 548 | |||
| AK166143.1 | - | 2, 3 | 535 | |||
| AK171310.1 | - | 2, 3 | 536 | |||
| AK172921.1 | - | 2, 3 | 590 | |||
| AK202332.1 | - | 1, 2 | 72 | |||
| AK204727.1 | - | 1, 2 | 72 | |||
| BC053030.1 | - | 2, 3 | 537 | |||
| BC059006.1 | - | 2, 3 | 531 | 6827203 | ||

Ip6k1
Genomic Map