siRNA Details
| Gene Symbol: Klra16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013794.4 | NP_038822.3 | 4 | 407 | ||
| GenBank mRNA | AF146570.4 | - | 4 | 407 | ||
| AY971807.1 | - | 3 | 305 | |||
| BC119242.1 | - | 4 | 396 | 8733991 | ||
| BC144924.1 | - | 4 | 396 | 9053472 | ||
| BC144940.1 | - | 4 | 434 | 9053488 | ||

Klra16
Genomic Map