siRNA Details
| Gene Symbol: MFSD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001120.4 | NP_001111.3 | 12 | 1925 | ||
| NM_001146069.1 | NP_001139541.1 | 13 | 1723 | |||
| XM_005247927.3 | XP_005247984.1 | 12 | 1825 | |||
| GenBank mRNA | BC001502.2 | - | 13 | 1649 | 2988084 | |
| BC014979.1 | - | 13 | 1652 | 4878602 | ||
| L11669.1 | - | 12 | 1731 | |||

MFSD10
Genomic Map