siRNA Details
| Gene Symbol: Mocs2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113374.1 | NP_001106845.1 | 2 | 217 | ||
| NM_001113375.1 | NP_001106846.1 | 2 | 217 | |||
| NM_013826.3 | NP_038854.2 | 3 | 339 | |||
| GenBank mRNA | AF091872.1 | - | 3 | 290 | ||
| AK002719.1 | - | 2 | 185 | |||
| AK010112.1 | - | 3 | 319 | |||
| AK078782.1 | - | 2 | 185 | |||
| CX220128.1 | - | 2 | 167 | |||

Mocs2
Genomic Map