siRNA Details
| Gene Symbol: Tcf20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114140.1 | NP_001107612.1 | 2 | 3402 | ||
| NM_013836.3 | NP_038864.3 | 2 | 3402 | |||
| GenBank mRNA | AK122239.1 | - | 1 | 2431 | ||
| AY007594.1 | - | 2 | 3402 | |||
| BC042485.1 | - | 2 | 781 | |||
| BC080855.1 | - | 1 | 528 | |||
| BC138038.1 | - | 2 | 3375 | 8861057 | ||
| BC171946.1 | - | 2 | 3375 | |||
| U20282.1 | - | 1 | 902 | |||

Tcf20
Genomic Map