siRNA Details
Gene Symbol: Tcl1b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013776.1 | NP_038804.1 | 4 | 882 | ||
GenBank mRNA | AF195493.1 | - | 4 | 882 | ||
AK139627.1 | - | 4 | 897 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tcl1b5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013776.1 | NP_038804.1 | 4 | 882 | ||
GenBank mRNA | AF195493.1 | - | 4 | 882 | ||
AK139627.1 | - | 4 | 897 |