siRNA Details
| Gene Symbol: Npas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013780.2 | NP_038808.2 | 3 | 299 | ||
| GenBank mRNA | AF173871.1 | - | 3 | 299 | ||
| BC120885.1 | - | 2 | 209 | |||
| BC148199.2 | - | 2 | 209 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Npas3| Gene Symbol: Npas3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013780.2 | NP_038808.2 | 3 | 299 | ||
| GenBank mRNA | AF173871.1 | - | 3 | 299 | ||
| BC120885.1 | - | 2 | 209 | |||
| BC148199.2 | - | 2 | 209 | |||