siRNA Details
Gene Symbol: Ptdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013782.4 | NP_038810.2 | 2, 3 | 344 | ||
GenBank mRNA | AF099053.1 | - | 2, 3 | 228 | ||
AK021249.1 | - | 2, 3 | 309 | |||
AK031906.1 | - | 2, 3 | 330 | |||
AK032686.1 | - | 2, 3 | 309 | |||
AK036368.1 | - | 2, 3 | 345 | |||
AK048026.1 | - | 2, 3 | 327 | |||
AK081275.1 | - | 2, 3 | 311 | |||
BC009013.1 | - | 11 | ||||
BC053517.1 | - | 2, 3 | 281 | 6315175 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106316, XM_006230532, BC166496.
Panther Classification:
Gene Ontology Categories:
