siRNA Details
| Gene Symbol: Ip6k1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013785.2 | NP_038813.2 | 3 | 554 | ||
| GenBank mRNA | AF177144.1 | - | 2 | 254 | ||
| AK088872.1 | - | 3 | 571 | |||
| AK166143.1 | - | 3 | 558 | |||
| AK171310.1 | - | 3 | 559 | |||
| AK172921.1 | - | 3 | 613 | |||
| AK180416.1 | - | 2 | 71 | |||
| AK184546.1 | - | 2 | 71 | |||
| AK202332.1 | - | 2 | 95 | |||
| AK204727.1 | - | 2 | 95 | |||
| BC059006.1 | - | 3 | 554 | 6827203 | ||

Ip6k1
Genomic Map