siRNA Details
| Gene Symbol: Klra15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013793.2 | NP_038821.2 | 6 | 1001 | ||
| GenBank mRNA | AF146571.2 | - | 6 | 1001 | ||
| BC125466.1 | - | 6 | 1053 | 40129981 | ||
| BC125494.1 | - | 6 | 1053 | 40130009 | ||

Klra15
Genomic Map