siRNA Details
| Gene Symbol: Klra16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013794.4 | NP_038822.3 | 7 | 1043 | ||
| GenBank mRNA | AF146570.4 | - | 7 | 1043 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klra16| Gene Symbol: Klra16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013794.4 | NP_038822.3 | 7 | 1043 | ||
| GenBank mRNA | AF146570.4 | - | 7 | 1043 | ||