siRNA Details
| Gene Symbol: Slco1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013797.5 | NP_038825.1 | 7 | 1160 | ||
| GenBank mRNA | AB031813.1 | - | 7 | 1001 | ||
| AF148218.1 | - | 7 | 984 | |||
| AF223067.1 | - | 7 | 994 | |||
| AK041125.1 | - | 7 | 1008 | |||
| AK165360.1 | - | 7 | 1015 | |||
| AY195868.1 | - | 7 | 993 | |||
| AY195869.1 | - | 7 | 993 | |||
| AY243579.1 | - | 7 | 993 | |||
| BC041147.1 | - | 7 | 943 | 4975986 | ||

Slco1a1
Genomic Map