siRNA Details
| Gene Symbol: Dbnl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146308.1 | NP_001139780.1 | 3 | 268 | ||
| NM_001146309.1 | NP_001139781.1 | 3 | 268 | |||
| NM_013810.3 | NP_038838.1 | 3 | 268 | |||
| GenBank mRNA | AK046073.1 | - | 3 | 248 | ||
| AK053796.1 | - | 3 | 264 | |||
| AK078082.1 | - | 3 | 235 | |||
| AK134136.1 | - | 3 | 235 | |||
| AK134487.1 | - | 3 | 235 | |||
| AK140752.1 | - | 3 | 235 | |||
| AK142818.1 | - | 3 | 218 | |||
| AK146920.1 | - | 3 | 235 | |||
| AK151096.1 | - | 3 | 248 | |||
| AK152110.1 | - | 3 | 233 | |||
| AK152204.1 | - | 3 | 248 | |||
| AK153072.1 | - | 3 | 248 | |||
| AK153389.1 | - | 3 | 222 | |||
| AK159984.1 | - | 3 | 245 | |||
| AK161973.1 | - | 3 | 248 | |||
| AK167956.1 | - | 3 | 208 | |||
| AK168898.1 | - | 3 | 245 | |||
| AK172471.1 | - | 3 | 225 | |||
| AK199448.1 | - | 3 | 145 | |||
| AY098595.1 | - | 3 | 191 | |||
| BC046430.1 | - | 3 | 233 | 4986658 | ||
| BY293844.1 | - | 3 | 268 | |||
| U58884.1 | - | 3 | 221 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_031352, NM_001277212, NM_001277213, NM_001277211, AB009346, and others.

Dbnl
Genomic Map