siRNA Details
| Gene Symbol: Azi2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286508.1 | NP_001273437.1 | 8 | 2293 | ||
| NM_013727.4 | NP_038755.1 | 8 | 2375 | |||
| GenBank mRNA | AB007141.1 | - | 8 | 2211 | ||
| AK030076.1 | - | 8 | 2207 | |||
| AK039794.1 | - | 1 | 492 | |||
| BC006851.1 | - | 8 | 2270 | 3156857 | ||

Azi2
Genomic Map