siRNA Details
| Gene Symbol: Phlda3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013750.2 | NP_038778.1 | 2 | 1410 | ||
| GenBank mRNA | AK003767.1 | - | 2 | 1376 | ||
| AK189460.1 | - | 1 | 106 | |||
| AK195224.1 | - | 1 | 106 | |||
| AK213926.1 | - | 106 | ||||
| AK214179.1 | - | 106 | ||||
| AK218195.1 | - | 1 | 137 | |||
| BC023376.1 | - | 2 | 1319 | |||
| BC023408.1 | - | 2 | 1319 | 5252055 | ||

Phlda3
Genomic Map