siRNA Details
| Gene Symbol: Cyp2g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013809.1 | NP_038837.1 | 9 | 1691 | ||
| GenBank mRNA | AK014160.1 | - | 9 | 1701 | ||
| BC046321.1 | - | 9 | 1726 | |||
| BC058222.1 | - | 9 | 1696 | 6306453 | ||
| L81171.1 | - | 9 | 1691 | |||

Cyp2g1
Genomic Map