siRNA Details
| Gene Symbol: Hk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013820.3 | NP_038848.1 | 18 | 4315 | ||
| GenBank mRNA | AJ238540.1 | - | 18 | 4258 | ||
| AK036845.1 | - | 1 | 455 | |||
| BB629860.1 | - | 1 | 455 | |||
| BC054472.1 | - | 18 | 4232 | 6465367 | ||

Hk2
Genomic Map