siRNA Details
| Gene Symbol: Hsd3b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013821.3 | NP_038849.2 | 4 | 1492 | ||
| GenBank mRNA | AF031170.1 | - | 3 | 1232 | ||
| AK142267.1 | - | 4 | 1494 | |||
| AY046512.1 | - | 4 | 1483 | |||
| BC053501.1 | - | 4 | 1478 | 30018625 | ||

Hsd3b6
Genomic Map