siRNA Details
| Gene Symbol: Vps45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013841.3 | NP_038869.1 | 1 | 758 | ||
| GenBank mRNA | AK083678.1 | - | 1 | 134 | ||
| AK146190.1 | - | 1 | 160 | |||
| AK168101.1 | - | 1 | 160 | |||
| BC012691.1 | - | 1 | 134 | 4038142 | ||
| BC058528.1 | - | 1 | 150 | 5718352 | ||
| U66865.1 | - | 1 | 773 | |||

Vps45
Genomic Map