siRNA Details
| Gene Symbol: Zfp385a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013866.2 | NP_038894.2 | 4 | 308 | ||
| GenBank mRNA | AF118566.1 | - | 4 | 306 | ||
| AK137748.1 | - | 4 | 315 | |||
| BC017644.1 | - | 4 | 365 | 4504518 | ||
| CK619761.1 | - | 4 | 306 | |||

Zfp385a
Genomic Map