siRNA Details
| Gene Symbol: Scmh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159630.1 | NP_001153102.1 | 9 | 1083 | ||
| NM_013883.2 | NP_038911.1 | 9 | 1083 | |||
| GenBank mRNA | AB030906.1 | - | 9 | 982 | ||
| AB221680.1 | - | 5 | 624 | |||
| AB221681.1 | - | 5 | 624 | |||
| BC034667.1 | - | 7 | 793 | 5356322 | ||

Scmh1
Genomic Map