siRNA Details
| Gene Symbol: Fbxw5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013908.4 | NP_038936.1 | 2 | 281 | ||
| GenBank mRNA | AF176520.1 | - | 2 | 242 | ||
| AK019715.2 | - | 2 | 282 | |||
| AK154911.1 | - | 1 | 1220 | |||
| BC010776.1 | - | 2 | 208 | 4211592 | ||

Fbxw5
Genomic Map