siRNA Details
| Gene Symbol: Pttg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131054.1 | NP_001124526.1 | 3 | 160 | ||
| NM_013917.2 | NP_038945.2 | 2 | 412 | |||
| GenBank mRNA | AF069051.1 | - | 2 | 404 | ||
| AF071209.1 | - | 2 | 412 | |||
| AK002473.1 | - | 2 | 126 | |||
| AK008704.1 | - | 2 | 127 | |||
| AK088387.1 | - | 2 | 126 | |||
| BC023324.1 | - | 3 | 166 | 4946234 | ||
| CN837307.1 | - | 3 | 165 | |||
| DQ157177.1 | - | 2 | 101 | |||
| DV657646.1 | - | 2 | 130 | |||

Pttg1
Genomic Map