siRNA Details
| Gene Symbol: Zfp68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044747.2 | NP_001038212.1 | 3 | 310 | ||
| NM_001163797.1 | NP_001157269.1 | 4 | 400 | |||
| NM_013844.2 | NP_038872.1 | 4 | 406 | |||
| GenBank mRNA | AB024005.1 | - | 4 | 390 | ||
| AK013868.1 | - | 4 | 401 | |||
| AK044831.1 | - | 3 | 280 | |||
| AK146092.1 | - | 4 | 367 | |||
| AK153887.1 | - | 4 | 342 | |||
| BC018171.1 | - | 4 | 389 | 3989444 | ||
| BY088951.1 | - | 3 | 313 | |||

Zfp68
Genomic Map