siRNA Details
| Gene Symbol: Ror2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013846.3 | NP_038874.3 | 2, 3 | 415 | ||
| GenBank mRNA | AB010384.1 | - | 2, 3 | 183 | ||
| AK031112.1 | - | 2, 3 | 385 | |||
| AK031789.1 | - | 2, 3 | 173 | |||
| AK083538.1 | - | 2, 3 | 391 | |||
| AK084752.1 | - | 2, 3 | 381 | |||
| AK157705.1 | - | 2, 3 | 431 | |||
| BC070436.1 | - | 2, 3 | 415 | 30535615 | ||

Ror2
Genomic Map