siRNA Details
| Gene Symbol: Hspb7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013868.4 | NP_038896.2 | 2 | 1228 | ||
| GenBank mRNA | AF155909.1 | - | 298 | |||
| AJ243192.1 | - | 298 | ||||
| AK030025.1 | - | 2 | 1229 | |||
| AK086011.1 | - | 2 | 305 | |||
| BC046993.1 | - | 2 | 264 | |||
| BC089584.1 | - | 2 | 303 | 6755510 | ||

Hspb7
Genomic Map