siRNA Details
| Gene Symbol: Rnf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013876.3 | NP_038904.1 | 2 | 476 | ||
| GenBank mRNA | AB024427.1 | - | 3 | 341 | ||
| AF220206.1 | - | 2 | 230 | |||
| AK014254.1 | - | 2 | 630 | |||
| AK183757.1 | - | 1 | 91 | |||
| BC010299.1 | - | 2 | 476 | 3155804 | ||
| BC028255.1 | - | 2 | 657 | 4972336 | ||

Rnf11
Genomic Map