siRNA Details
| Gene Symbol: Scmh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159630.1 | NP_001153102.1 | 10, 11 | 1414 | ||
| NM_013883.2 | NP_038911.1 | 10, 11 | 1414 | |||
| GenBank mRNA | AB030906.1 | - | 10, 11 | 1313 | ||
| AB221680.1 | - | 6, 7 | 955 | |||
| AB221681.1 | - | 6, 7 | 955 | |||
| BC025478.1 | - | 1, 2 | 1656 | |||
| BC034667.1 | - | 8, 9 | 1124 | 5356322 | ||

Scmh1
Genomic Map