siRNA Details
| Gene Symbol: Dnajc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253685.1 | NP_001240614.1 | 3 | 195 | ||
| NM_013888.3 | NP_038916.1 | 3 | 332 | |||
| GenBank mRNA | AF132906.2 | - | 3 | 281 | ||
| AK002649.1 | - | 1 | 20 | |||
| AK032207.1 | - | 2 | 1701 | |||
| BC013501.1 | - | 3 | 239 | 4222828 | ||
| BC071211.1 | - | 2 | 1597 | |||
| BF453706.1 | - | 3 | 195 | |||
| DV042983.1 | - | 3 | 347 | |||

Dnajc12
Genomic Map