siRNA Details
| Gene Symbol: Reg3d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161741.1 | NP_001155213.1 | 4, 5 | 387 | ||
| NM_013893.2 | NP_038921.2 | 5, 6 | 525 | |||
| GenBank mRNA | AB028625.1 | - | 5, 6 | 531 | ||
| AB035204.1 | - | 5, 6 | 505 | |||
| AK019033.1 | - | 5, 6 | 504 | |||
| BC113760.1 | - | 5, 6 | 520 | 40055351 | ||
| BM055959.1 | - | 3, 4 | 335 | |||

Reg3d
Genomic Map