siRNA Details
| Gene Symbol: Fbxl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013909.2 | NP_038937.2 | 2 | 542 | ||
| GenBank mRNA | AB041571.1 | - | 1 | 780 | ||
| AF176522.1 | - | 2 | 530 | |||
| AK040458.1 | - | 1 | 792 | |||
| AK052585.1 | - | 1 | 781 | |||
| AK177913.1 | - | 1 | 31 | |||
| AK183822.1 | - | 1 | 32 | |||
| BC011485.1 | - | 1 | 785 | |||
| BC026818.1 | - | 1 | 779 | |||
| BC044784.2 | - | 1 | 742 | |||
| BC054377.1 | - | 2 | 519 | 6315438 | ||

Fbxl6
Genomic Map