siRNA Details
| Gene Symbol: Fbxl12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001002846.2 | NP_001002846.1 | 3 | 1575 | ||
| NM_001286529.1 | NP_001273458.1 | 3 | 1197 | |||
| NM_001286530.1 | NP_001273459.1 | 4 | 1301 | |||
| NM_013911.3 | NP_038939.2 | 3 | 1605 | |||
| GenBank mRNA | AF176525.1 | - | 3 | 1486 | ||
| AK011081.1 | - | 3 | 1560 | |||
| AK012811.1 | - | 3 | 1098 | |||
| AK029429.1 | - | 1 | 540 | |||
| AK030910.1 | - | 3 | 1575 | |||
| AK031262.1 | - | 2 | 3661 | |||
| AK137282.1 | - | 4 | 3272 | |||

Fbxl12
Genomic Map