siRNA Details
| Gene Symbol: Rlim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011276.3 | NP_035406.3 | 5 | 4465 | ||
| GenBank mRNA | AF197105.1 | - | 1 | 589 | ||
| AK052543.1 | - | 1 | 745 | |||
| BC053711.1 | - | 1 | 477 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rlim| Gene Symbol: Rlim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011276.3 | NP_035406.3 | 5 | 4465 | ||
| GenBank mRNA | AF197105.1 | - | 1 | 589 | ||
| AK052543.1 | - | 1 | 745 | |||
| BC053711.1 | - | 1 | 477 | |||