siRNA Details
| Gene Symbol: Siva1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013929.2 | NP_038957.2 | 2 | 351 | ||
| GenBank mRNA | AF033113.1 | - | 2 | 212 | ||
| AF033114.1 | - | 2 | 299 | |||
| AK010562.1 | - | 2 | 277 | |||
| AK017570.1 | - | 2 | 312 | |||
| AK153115.1 | - | 2 | 261 | |||
| AK169200.1 | - | 2 | 304 | |||
| AK184852.1 | - | 2 | 156 | |||
| AK209128.1 | - | 2 | 156 | |||
| AK212226.1 | - | 2 | 156 | |||
| BC012279.1 | - | 2 | 333 | 4013026 | ||

Siva1
Genomic Map