siRNA Details
| Gene Symbol: Pttg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013917.2 | NP_038945.2 | 4 | 1189 | ||
| GenBank mRNA | AF071209.1 | - | 4 | 1189 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pttg1| Gene Symbol: Pttg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013917.2 | NP_038945.2 | 4 | 1189 | ||
| GenBank mRNA | AF071209.1 | - | 4 | 1189 | ||